Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

OmniTop Assemblies, Pre-Sterilized Single-Use Sample Tubes®, PAW BioScience

Supplier: PAW BioScience, VWR Bioprocessing Division
15CTAPPCFXST 15CTAPPCFDSLST 15CTAPPCFX00 15CTAPPCFDSL00 50CTA-CF-X-ST 50CTA-CF-DSLST 50CTA-CF-X-00 15CTAPSCFDSL00 15CTAPSCFDSLST 15CTAPSCFXST 15CTAPSCFX00 50CTA-CF-DSL00
OmniTop Sample Tubes® are a convenient device that can be used to obtain fluid samples. Each OmniTop tube comes with a pre-attached 0.2µm vent filter and 18" of tubing (C-Flex®, TYGON®, silicone or PharMed®).

  • Customizable cap system
  • Unique cap design allows for complete customization
  • Ability to use different ID and OD tubing diameters
  • Wide selection of tubing materials
  • Easily configured with virtually any type of tubing
  • Single-use
  • Cost effective
  • Reduced assembly and installation time
  • Eliminate cleaning validation
  • Flexible manufacturing
  • Design permits quick delivery on small lots
  • Closure system can be used with various glass bottles
  • Available individually packaged or configured in manifold

These tubes are available with or without internal dip-tubes to facilitate removal of the fluid inside

OmniTop Sample Tubes® can be aseptically connected to your bioreactor or other device using standard tubing welders, such as the Terumo SCD-IIB. Sample Tubes can also be aseptically sealed and removed (wet or dry) using tubing sealers such as the Genesis SE340 or handheld battery powered version SE640.

Use OmniTop for the transfer of biopharmaceutical products and reagents in a closed system, gas exchange transfer, sampling of biopharmaceutical products and reagents in a closed system, storage of biopharmaceutical products and reagents. For use with bench top, pilot plant, and production bioreactors. Also for use with sterile connecting devices and thermal or RF tubing sealers.

OmniTop Sample Tubes® are availble in a wide variety of standard configurations or they can be customized to suit your specific applications.




 

ORDER

Expand All / Collapse All

OmniTop 15 mL Sample Tubes
Description Material Capacity Supplier No. VWR Catalog Number Unit Price Quantity
OmniTop Assembly, C-Flex®, Gamma Polypropylene 15 mL 15CTAPPCFX00 75840-758 Case of 50 Retrieving
Each Retrieving
Pack of 10 Retrieving
OmniTop Assembly, C-Flex®, Silicone Dip Tube, Gamma Polypropylene 15 mL 15CTAPPCFDSL00 75840-754 Case of 50 Retrieving
Each Retrieving
Pack of 10 Retrieving
OmniTop Assembly, C-Flex®, Seal and Trim, Gamma Polypropylene 15 mL 15CTAPPCFXST 75840-760 Case of 50 Retrieving
Each Retrieving
Pack of 10 Retrieving
OmniTop Assembly, C-Flex®, Silicone Dip Tube, Seal and Trim, Gamma Polypropylene 15 mL 15CTAPPCFDSLST 75840-756 Case of 50 Retrieving
Each Retrieving
Pack of 10 Retrieving
OmniTop Assembly, C-Flex®, Gamma Polystyrene 15 mL 15CTAPSCFX00 75840-766 Case of 50 Retrieving
Each Retrieving
Pack of 10 Retrieving
OmniTop Assembly, C-Flex®, Silicone Dip Tube, Gamma Polystyrene 15 mL 15CTAPSCFDSL00 75840-762 Case of 50 Retrieving
Each Retrieving
Pack of 10 Retrieving
OmniTop Assembly, C-Flex®, Seal and Trim, Gamma Polystyrene 15 mL 15CTAPSCFXST 75840-768 Case of 50 Retrieving
Each Retrieving
Pack of 10 Retrieving
OmniTop Assembly, C-Flex®, Silicone Dip Tube, Seal and Trim, Gamma Polystyrene 15 mL 15CTAPSCFDSLST 75840-764 Case of 50 Retrieving
Each Retrieving
Pack of 10 Retrieving

OmniTop 50 mL Sample Tubes
Description Material Capacity Supplier No. VWR Catalog Number Unit Price Quantity
OmniTop Assembly, C-Flex®, Gamma Polypropylene 50 mL 50CTA-CF-X-00 75840-774 Case of 50 Retrieving
Each Retrieving
Pack of 10 Retrieving
OmniTop Assembly, C-Flex®, Silicone Dip Tube, Gamma Polypropylene 50 mL 50CTA-CF-DSL00 75840-770 Case of 50 Retrieving
Each Retrieving
Pack of 10 Retrieving
OmniTop Assembly, C-Flex®, Seal and Trim, Gamma Polypropylene 50 mL 50CTA-CF-X-ST 75840-776 Case of 50 Retrieving
Each Retrieving
Pack of 10 Retrieving
OmniTop Assembly, C-Flex®, Silicone Dip Tube, Seal and Trim, Gamma Polypropylene 50 mL 50CTA-CF-DSLST 75840-772 Case of 50 Retrieving
Each Retrieving
Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping