Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Gas Leak Detection Systems

https://ca.vwr.com/stibo/smallweb/std.lang.all/52/19/21655219.jpg

Compact and convenient, these sensitive gas leak detection systems locate the smallest leaks with functions such as LED features and one-hand operation. VWR also features gas leak detectors with three-color leak indication; visible and audible gas-concentration alarms; and thumb-controlled adjustable sensitivity to indicate hydrogen, methane, and other dangerous gases. For specific detection needs, VWR provides instruments that indicate at 10% (LEL) Lower Explosive Limity for methane. Both battery-run gas leak detection systems with batteries and detectors with chargers are available.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"2"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  Thermo Scientific
Description:   The Thermo Scientific GLD Gas Leak Detector is specifically designed for use with gas chromatography instruments.
Catalog Number: (97015-599)
Supplier:  Restek
Description:   Backed by a one-year warranty, the Restek leak detector is the industry standard for performance and affordability in handheld leak detectors.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 2  of 2