Would you like to log in again?
Preservation Plate, Watson Bio Lab
Supplier: Watson Bio Lab
176-502C 176-501C 176-302C 176-301C 176-201C 176-202C 176-104GR 176-103GR
Watson Preservation Plates store and enable transportation of such bio resources as nucleic acids (DNA, RNA, and oligonucleotide) and blood under room temperature and pressure.
  • Space Saving
  • Plates Prevent Sample Deterioration
  • No Reagent or Salt Used
  • Low Contamination Risk
  • Simple Storage Management
Marking space on the plate and a compact body make sample storage management easier.
Reagent and salt-free paper chips can be put directly into the solution to start PCR or in situ hybridization. A Preservation Plate (PVP) can reduce contamination risk by using different wells on the PVP or changing the PVP sheet itself, dependent on the sample.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Material Supplier No. VWR Catalog Number Unit Price Quantity
1-Well Preservation Plate Nylon 176-202C 75983-012 Box of 10 Retrieving
Each Retrieving
1-Well Preservation Plate Cellulose 176-201C 75983-010 Box of 10 Retrieving
Each Retrieving
3-Well Preservation Plate Nylon 176-302C 75983-008 Box of 10 Retrieving
Each Retrieving
3-Well Preservation Plate Cellulose 176-301C 75983-006 Box of 10 Retrieving
Each Retrieving
24-Well Preservation Plate Nylon 176-104GR 75983-004 Box of 5 Retrieving
Each Retrieving
24-Well Preservation Plate Cellulose 176-103GR 75983-002 Box of 5 Retrieving
Each Retrieving
96-Well Preservation Plate Nylon 176-502C 75983-000 Box of 5 Retrieving
Each Retrieving
96-Well Preservation Plate Cellulose 176-501C 75982-998 Box of 5 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty