Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ward's® Algae Beads (Scenedesmus sp.)

Supplier: VWR
Ward's®
WARD470225-514 SS0274-500ML
Demonstrate photosynthesis and respiration in action with our ready-made algae beads.

  • Let us do the class preparation for you!
  • Our beads are hardy and hold up well to handling by the most inexperienced students
  • Algae balls last at least 1 month under correct storage conditions
  • No complicated media needed, store in deionized or spring water

Simply place beads in Bicarbonate Indicator (also available) and watch the color change as beads move from dark to light environments. Students will love this powerful demonstration of the switch from respiration to photosynthesis, and come up with different ways to speed/slow or even reverse the reaction.

For maximum freshness, place beads in the refrigerator upon receipt. Beads can be reused once or twice by simply rinsing and returning them to the storage jar with fresh deionized or spring water (do not use distilled water)

Ordering information: These products contain living or perishable materials. Please provide a delivery date at checkout.



 

ORDER

Expand All / Collapse All
Description No. of Lab Groups Supplier No. VWR Catalog Number Unit Price Quantity
Algae Beads 8   470225-514 Each Retrieving
Pack of 200 Retrieving
Bicarbonate Indicator (500 mL) SS0274-500ML 470225-836 Each (500ml) Retrieving Restricted
restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty