Would you like to log in again?
Ward's® Essentials The Ins and Outs of Diffusion Kit
Supplier: VWR
Ward's®
How big can a cell grow?
  • Hands on lab activity
  • Kit Aligned to Next Generation Science Standard MS-LS1-2
  • Instructions and materials included for 15 groups
  • Middle school
Cell size is determined by how easily a cell can get nutrients and excrete its wastes. Using agar cubes as a model for a cell, students will determine the best cell size to allow waste elimination and nutrient acquisition.
Ordering information: Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, email or online.
ORDER
Expand All / Collapse All
Description No. of Lab Groups Supplier No. VWR Catalog Number Unit Price Quantity
The Ins and Outs of Diffusion Kit 15   470227-162 Kit of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty