Would you like to log in again?
Cell Counter Plates, Watson Bio Lab
Supplier: Watson Bio Lab
177-212C 177-112C 177-512C 177-312C
The ready-mounted plastic cover film provides easy handling.
  • Better Grid Linearity and Smoothness Compared to Glass Plates
  • Less Expensive than Glass Plates
  • Clear Visibility Makes Counting Easy
  • Ready-Mounted Plastic Cover Film on Plates
  • Four-Grid Patterns Are Available
It's easy to count the grid lines molded in the groove form because cells don't get stuck. Cells are easy to observe because the grid lines are thinner and smoother than glass slides.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Burker-Turk Cell Counting Plate 177-212C 75982-982 Each Retrieving
Pack of 10 Retrieving
Fuchs Rosenthal Cell Counting Plate 177-512C 75982-986 Each Retrieving
Pack of 10 Retrieving
Improved Neubauer Cell Counting Plate 177-112C 75982-980 Each Retrieving
Pack of 10 Retrieving
Thoma Cell Counting Plate 177-312C 75982-984 Each Retrieving
Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty