2-(4-Thiazolyl)benzimidazole ≥98.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: Thiabendazole

T1076-100GM T1076-100GM T1076-500GM
CA95036-256EA 614.36 CAD
95036-256 CA95036-256 95036-260
2-(4-Thiazolyl)benzimidazole ≥98.0% USP
2-(4-Thiazolyl)benzimidazole

Thiabendazole, USP is an antihelmintic deworming drug. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₀H₇N₃S
Storage Temperature: Ambient
MDL Number: MFCD00005587
CAS Number: 148-79-8

Order Now

Specification Test Results

Assay (Dried Basis) 98.0-101.0 %
Melting Range 296 - 303 °C
Chromatographic Purity To Pass Test
Loss On Drying max. 0.5 %
Residue On Ignition max. 0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR