Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

You Searched For:

Conductivity Sensors

23  results were found
https://ca.vwr.com/stibo/smallweb/std.lang.all/93/35/18989335.jpg

Conductivity sensors are becoming increasingly important for the determination of impurities in water or the concentration measurement of dissolved chemicals. Different models meet a variety of specific functions. Many of these sensors are intended for low-conductivity applications typically found in the semi-conductor, power, water, and pharmaceutical industries, providing accurate measurements even under extreme conditions in a conveniently compact instrument. Designs facilitate shielding during immersion measurements in various aqueous and non-aqueous media.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"23"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   VWR® electrodes offer exceptional response time and accuracy.
Product available on GSA Advantage®
Supplier:  Hach
Description:   IntelliCAL CDC401 is a digital, graphite, 4-pole conductivity probe
Supplier:  Hach
Description:   The IntelliCAL™ CDC401 is a digital, graphite, 4-pole conductivity cell with a temperature sensor
Catalog Number: (CA89501-070)

Supplier:  Hach
Description:   Hach offers conductivity cells for a wide variety of applications
Supplier:  Hach
Description:   The Radiometer CDC511T is a 4-pole conductivity cell with non-platinized, platinum sensor (k = 1.0 cm-1), integrated temperature sensor and TPX body
Supplier:  Hach
Description:   The Radiometer Analytical CDC641T conductivity cell is a 2-pole conductivity cell with platinum sensor
Supplier:  Hach
Description:   The Radiometer CDC861T is a 4-pole conductivity cell with platinized, platinum sensor (k = 1.0 cm-1), integrated temperature sensor and glass body
Supplier:  Hach
Description:   The Radiometer CDC741T is a 2-pole conductivity cell with platinized, platinum sensor (k = 1.0 cm-1), glass body and integrated temperature sensor
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 23
  1  2  3  Next