Order Entry
ContactUsLinkComponent
Cell-Mate3D™ RNA Isolation Kit, BRTI Life Sciences
Cell-Mate3D™ RNA Isolation Kit, BRTI Life Sciences
Catalog #: 76036-002
Supplier:  Avantor
Cell-Mate3D™ RNA Isolation Kit, BRTI Life Sciences
Catalog #: 76036-002
Supplier:  Avantor
Discontinued
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Method/Format:
    Enzymatic extraction
  • Prep size:
    Scalable
  • Refine target molecule:
    Total RNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Size:
    30 Reactions
  • Target molecule:
    RNA
  • Description:
    Cell-Mate3D™ RNA Isolation Kit without Beads
  • Description:
    Cell-Mate3D™ RNA Isolation Kit without Beads

Specifications

About this item

Isolation RNA from Cell-Mate3D matrix for cancer cells, stem cells, or tissue engineering.

  • Isolate RNA from polysaccharide rich matrix
  • 30 reactions per kit
  • 2 year shelf-life on kit
  • Perform q-RT PCR
Customers Who Bought This Also Bought