Masterflex® I/P® Stainless-Steel IP66 Digital Dispensing Pump System, Cole-Parmer
Supplier: Cole Parmer
77420-18 77715-26
76047-520EA 14097.23 CAD
76047-520 76047-508
Masterflex® I/P® Stainless-Steel IP66 Digital Dispensing Pump System, Cole-Parmer
Pumps Peristaltic Pumps
Dispense by Volume in mL (0.001 to 99999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle).
  • Graphical LCD shows four operating modes continuous run, timed dispense, copy dispense, volume dispense
  • View pump performance continually
  • Antidrip function ensures dispensing accuracy
  • Open head sensor
These precision drives offer analog remote control options and simple programming—ideal for automated process applications. Drives feature a programmable dispense interval that lets you set the delay between dispense cycles for convenient, automated dispensing. Programmed calibration ensures dispense and display accuracy—drive stores one calibration value per tubing size.
Program the drive via the simple membrane keypad; keypad lock/unlock feature protects settings. Motor is reversible so you can purge tubing before or after pumping and pump fluid in either direction
Certifications: ETL, cETL, CE marked.
Ordering information: Includes NEMA 3-prong ground power cable. Accessories not included.