Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Avanti™ J-15 Series Benchtop Centrifuges, Beckman Coulter®

Supplier: BECKMAN COULTER CANADA LP CA
Avanti™
C01994 C01995 C19420 C19400 C19422 C19389 C19421 C19424 C19402 C19423 C19401 C19404 C19403 C19406 C19405 C19408 C19407 C19409 C19390 C19392 C19391 C19394 C19393 B99517 C19396 B99516 C19395 C19398 C19397 C19411 C19399 C19410 B99515 C19413 B99514 C19412 C19415 C19414 C19417 C19416 C19419 C19418
The new Avanti™ J-15 Series of benchtop centrifuge (refrigerated or ventilated) leverages the Ultra Harmonic Technology, designed to protect your sample and increase workflow time efficiencies.

  • Ultra harmonic technology
  • Workflow optimization
  • Sample protection
  • Time efficiency
  • Multi-dimension imbalance tolerance

The Avanti™ J-15 Series of benchtop centrifuges is part of a new family of life science equipment that provides the exceptional performance you expect from Beckman Coulter

Refrigerated (Avanti™ J-15R) or ventilated (Avanti™ J-15), these centrifuges use Ultra Harmonic Technology. Ultra Harmonic Technology leverages intelligent acceleration and deceleration profiles, reduces sample disturbances and provides time savings.

The Avanti™ J-15 series is an all around solution for your scientific needs: cell culture, transfection/transduction, nucleic acid purification, blood processing, assay set-up and more. Routine cell culture procedures are made easy and safe with the use of aerosolve canister. These optional BioCertified* aerosolve canisters are made of clear shell to allow sample
visibility even before the canister is opened.

Speed Efficiency and Minimum Sample Disturbances with Ultra Harmonic Technology (intelligent acceleration and deceleration profiles).

Sample Protection and Higher Yield: soft pellet deceleration profile protects the sample and results in increased sample yield.
Optimize Bench Space (with our truly ventilated unit while processing any protocol in cell culture, blood separation or microbiology).
Ease of Use and Faster Accessibility: icon usability and intuitive display on high contrast color LCD screen.
Efficiency: Easy one-touch access to 6 programs and store up to 99 programs.
JS-4.750 swinging bucket rotor. (Spin from 1.5mL reaction vials to 750 mL bottles, single to quad-pack blood bags and multi-well plates, with the proper adapters or microplate carriers.)

JA-10.100 (6 x 100 mL) BioCertified* fixed angle rotor. (Spin up to six 100 mL tubes at a 25° angle to the axis of rotation. The dual- locking lid allows the rotor to be removed from the centrifuge with the lid on.)
Applications for the JA-10.100 rotor include: density gradient separations of erythrocytes, cell lysate fractions, granules, as well as differential separation of DNA, proteins, and viruses. Up to 600 mL of sample volume can be centrifuged per run. Optional adapters allow spinning 1.5 microcentrifuge tubes, 10 mL round, 15 mL round and conical and 50 mL round and conical tubes.
*BioCertified: is a term used to describe our products which have been tested and validated to demonstrate containment of microbiological aerosols by an independent, third-party facility. Improper use or maintenance may affect seal integrity and , this, containment. BioSafe and BioSafety are terms intended to describe the enhanced biocontainment features of our products.
*Up to three Beckman Coulter plates can be run per carrier at maximum multi-well plate speeds. If running four Beckman Coulter plates, reduce run speed to 2,700 RPM. Observe manufacturer’s run speed and stacking limitations if using other manufacturers’ multi-well plates.
**Up to three Beckman Coulter plates, separated by cap strips, can be run per carrier at maximum multi-well plate speeds. If running four Beckman Coulter plates, cap strips cannot be used due to height limitations; reduce run speed to 2,700 RPM. Observe manufacturer’s run speed and stacking limitations if using other manufacturers’ multi-well plates.

Certifications: Meets ASTM standards. EN-342.

Ordering information: The centrifuge is supplied with a one-year parts and labor warranty. Rotors, adapters and accessories are sold separately.




 

ORDER

Expand All / Collapse All

Instruments Only
Description Electrical Supplier No. VWR Catalog Number Unit Price Quantity
Avanti™ J-15 Ventilated Benchtop Centrifuge, IVD 100 – 120 V AC, 50/60 Hz C01994 CA76121-334 Each Retrieving
Avanti™ J-15 Ventilated Benchtop Centrifuge, NON-IVD 100 – 120 V, 50/60 Hz C01995 CA76121-336 Each Retrieving

Centrifuge Packages
Description Electrical Supplier No. VWR Catalog Number Unit Price Quantity
Avanti™ J-15R Refrigerated Benchtop Centrifuge, IVD 120 V, 60 Hz B99517 CA76121-338 Each Retrieving
Avanti™ J-15R Refrigerated Benchtop Centrifge, NON-IVD 120 V, 60 Hz B99516 CA76121-314 Each Retrieving

Centrifuge Packages, 100 – 120 V
Description Electrical Includes Supplier No. VWR Catalog Number Unit Price Quantity
Avanti J-15, IVD, Ventilated, Benchtop Centrifuge Fixed Angle Package 100 – 120 V, 50/60 Hz Includes benchtop centrifuge and BioCertified JA-10.100 (6 x 100 mL) fixed angle rotor. Adapters, tubes/bottles are NOT included and can be ordered separately. C19393 CABKC19393 Each Retrieving
Avanti J-15, Ventilated, Benchtop Centrifuge Fixed Angle Package 100 – 120 V, 50/60 Hz Includes benchtop centrifuge and BioCertified JA-10.100 (6 x 100 mL) fixed angle rotor. Adapters, tubes/bottles are NOT included and can be ordered separately. C19394 CABKC19394 Each Retrieving
Avanti J-15, IVD, Ventilated, Benchtop Centrifuge Swinging Bucket Package 100 – 120 V, 50/60 Hz Includes benchtop centrifuge and BioCertified JS-4.750 (4 x 750 mL) swinging bucket rotor. (Rotor is BioCertified when used with bucket covers or aerosolve canisters.) Adapters, tubes/bottles, bucket covers and aerosolve canisters are NOT included and can be ordered separately. C19399 CABKC19399 Each Retrieving
Avanti J-15, Ventilated, Benchtop Centrifuge Swinging Bucket Package 100 – 120 V, 50/60 Hz Includes benchtop centrifuge and BioCertified JS-4.750 (4 x 750 mL) swinging bucket rotor. (Rotor is BioCertified when used with bucket covers or aerosolve canisters.) Adapters, tubes/bottles, bucket covers and aerosolve canisters are NOT included and can be ordered separately. C19400 CABKC19400 Each Retrieving
Avanti J-15, IVD,Ventilated, Benchtop Centrifuge Microplate Package 100 – 120 V, 50/60 Hz Includes benchtop centrifuge and JS-4.750μ microplate rotor (yoke with microplate carriers). Microplates and microplate covers are NOT included and can be ordered separately. C19405 CABKC19405 Each Retrieving
Avanti J-15, Ventilated, Refrigerated Benchtop Microplate Package 100 – 120 V, 50/60 Hz Includes benchtop centrifuge and JS-4.750μ microplate rotor (yoke with microplate carriers). Microplates and microplate covers are NOT included and can be ordered separately. C19406 CABKC19406 Each Retrieving
Avanti J-15, IVD, Ventilated, Benchtop Centrifuge BioSafe Package 100 – 120 V, 50/60 Hz Includes benchtop centrifuge and BioCertified S-4.750 (4 x 750 mL) swinging bucket rotor, BioCertified aerosolve canisters (qty 4), 15 mL conical BioSafe aerosolve canister tube racks (qty 4) and 50 mL conical BioSafe aerosolve canister tube racks (qty 4). (Rotor is BioCertified when used with aerosolve canisters.) Tubes/bottles are NOT included and can be ordered separately. C19411 CABKC19411 Each Retrieving
Avanti J-15, Ventilated, Benchtop Centrifuge BioSafe Package 100 – 120 V, 50/60 Hz Includes benchtop centrifuge and BioCertified S-4.750 (4 x 750 mL) swinging bucket rotor, BioCertified aerosolve canisters (qty 4), 15 mL conical BioSafe aerosolve canister tube racks (qty 4) and 50 mL conical BioSafe aerosolve canister tube racks (qty 4). (Rotor is BioCertified when used with aerosolve canisters.) Tubes/bottles are NOT included and can be ordered separately. C19412 CABKC19412 Each Retrieving
Avanti J-15, IVD,Ventilated, Benchtop Centrifuge Cell Culture Package 100 – 120 V, 50/60 Hz Includes benchtop centrifuge and BioCertified JS-4.750 (4 x 750 mL) swinging bucket rotor, 15 mL conical tube adapters (qty 4) and 50 mL conical tube adapters (qty 4). (Rotor is BioCertified when used with bucket covers.) Bucket covers and tubes/bottles are NOT included and can be ordered separately. C19417 CABKC19417 Each Retrieving
Avanti J-15, Ventilated, Benchtop Centrifuge Cell Culture Package 100 – 120 V, 50/60 Hz Includes benchtop centrifuge and BioCertified JS-4.750 (4 x 750 mL) swinging bucket rotor, 15 mL conical tube adapters (qty 4) and 50 mL conical tube adapters (qty 4). (Rotor is BioCertified when used with bucket covers.) Bucket covers and tubes/bottles are NOT included and can be ordered separately. C19418 CABKC19418 Each Retrieving
Avanti J-15, IVD,Ventilated, Benchtop Centrifuge Blood Sample Package 100 – 120 V, 50/60 Hz Includes benchtop centrifuge and BioCertified JS-4.750 (4 x 750 mL) swinging bucket rotor, 13 mm diameter tube adapters (qty 4) and 16 mm diameter tube adapters (qty 4). (Rotor is BioCertified when used with bucket covers.) Bucket covers and tubes/bottles are NOT included and can be ordered separately. C19423 CABKC19423 Each Retrieving
Avanti J-15, Ventilated, Benchtop Centrifuge Blood Sample Package 100 – 120 V, 50/60 Hz Includes benchtop centrifuge and BioCertified JS-4.750 (4 x 750 mL) swinging bucket rotor, 13 mm diameter tube adapters (qty 4) and 16 mm diameter tube adapters (qty 4). (Rotor is BioCertified when used with bucket covers.) Bucket covers and tubes/bottles are NOT included and can be ordered separately. C19424 CABKC19424 Each Retrieving

Centrifuge Packages, 120 V
Description Electrical Includes Supplier No. VWR Catalog Number Unit Price Quantity
Avanti J-15R, Refrigerated, Benchtop Centrifuge Fixed Angle Package 120 V, 60 Hz Includes benchtop centrifuge and BioCertified JA-10.100 (6 x 100 mL) fixed angle rotor. Adapters, tubes/bottles are NOT included and can be ordered separately. C19391 CABKC19391 Each Retrieving
Avanti J-15R, IVD, Refrigerated, Benchtop Centrifuge Fixed Angle Package 120 V, 60 Hz Includes benchtop centrifuge and BioCertified JA-10.100 (6 x 100 mL) fixed angle rotor. Adapters, tubes/bottles are NOT included and can be ordered separately. C19392 CABKC19392 Each Retrieving
Avanti J-15R, Refrigerated, Benchtop Centrifuge Swinging Bucket Package 120 V, 60 Hz Includes benchtop centrifuge and BioCertified JS-4.750 (4 x 750 mL) swinging bucket rotor. (Rotor is BioCertified when used with bucket covers or aerosolve canisters.) Adapters, tubes/bottles, bucket covers and aerosolve canisters are NOT included and can be ordered separately. C19397 CABKC19397 Each Retrieving
Avanti J-15R, IVD, Refrigerated, Benchtop Centrifuge Swinging Bucket Package 120 V, 60 Hz Includes benchtop centrifuge and BioCertified JS-4.750 (4 x 750 mL) swinging bucket rotor. (Rotor is BioCertified when used with bucket covers or aerosolve canisters.) Adapters, tubes/bottles, bucket covers and aerosolve canisters are NOT included and can be ordered separately. C19398 CABKC19398 Each Retrieving
Avanti J-15R, Refrigerated, Benchtop Centrifuge Microplate Package 120 V, 60 Hz Includes benchtop centrifuge and JS-4.750μ microplate rotor (yoke with microplate carriers). Microplates and microplate covers are NOT included and can be ordered separately. C19403 CABKC19403 Each Retrieving
Avanti J-15R, IVD,Refrigerated, Benchtop Centrifuge Microplate Package 120 V, 60 Hz Includes benchtop centrifuge and JS-4.750μ microplate rotor (yoke with microplate carriers). Microplates and microplate covers are NOT included and can be ordered separately. C19404 CABKC19404 Each Retrieving
Avanti J-15R, Refrigerated, Benchtop Centrifuge BioSafe Package 120 V, 60 Hz Includes benchtop centrifuge and BioCertified S-4.750 (4 x 750 mL) swinging bucket rotor, BioCertified aerosolve canisters (qty 4), 15 mL conical BioSafe aerosolve canister tube racks (qty 4) and 50 mL conical BioSafe aerosolve canister tube racks (qty 4). (Rotor is BioCertified when used with aerosolve canisters.) Tubes/bottles are NOT included and can be ordered separately. C19409 CABKC19409 Each Retrieving
Avanti J-15R, IVD, Refrigerated, Benchtop Centrifuge BioSafe Package 120 V, 60 Hz Includes benchtop centrifuge and BioCertified S-4.750 (4 x 750 mL) swinging bucket rotor, BioCertified aerosolve canisters (qty 4), 15 mL conical BioSafe aerosolve canister tube racks (qty 4) and 50 mL conical BioSafe aerosolve canister tube racks (qty 4). (Rotor is BioCertified when used with aerosolve canisters.) Tubes/bottles are NOT included and can be ordered separately. C19410 CABKC19410 Each Retrieving
Avanti J-15R, Refrigerated, Benchtop Centrifuge Cell Culture Package 120 V, 60 Hz Includes benchtop centrifuge and BioCertified JS-4.750 (4 x 750 mL) swinging bucket rotor, 15 mL conical tube adapters (qty 4) and 50 mL conical tube adapters (qty 4). (Rotor is BioCertified when used with bucket covers.) Bucket covers and tubes/bottles are NOT included and can be ordered separately. C19415 CABKC19415 Each Retrieving
Avanti J-15R, IVD, Refrigerated, Benchtop Centrifuge Cell Culture Package 120 V, 60 Hz Includes benchtop centrifuge and BioCertified JS-4.750 (4 x 750 mL) swinging bucket rotor, 15 mL conical tube adapters (qty 4) and 50 mL conical tube adapters (qty 4). (Rotor is BioCertified when used with bucket covers.) Bucket covers and tubes/bottles are NOT included and can be ordered separately. C19416 CABKC19416 Each Retrieving
Avanti J-15R, Refrigerated, Benchtop Centrifuge Blood Sample Package 120 V, 60 Hz Includes benchtop centrifuge and BioCertified JS-4.750 (4 x 750 mL) swinging bucket rotor, 13 mm diameter tube adapters (qty 4) and 16 mm diameter tube adapters (qty 4). (Rotor is BioCertified when used with bucket covers.) Bucket covers and tubes/bottles are NOT included and can be ordered separately. C19421 CABKC19421 Each Retrieving
Avanti J-15R, IVD, Refrigerated, Benchtop Centrifuge Blood Sample Package 120 V, 60 Hz Includes benchtop centrifuge and BioCertified JS-4.750 (4 x 750 mL) swinging bucket rotor, 13 mm diameter tube adapters (qty 4) and 16 mm diameter tube adapters (qty 4). (Rotor is BioCertified when used with bucket covers.) Bucket covers and tubes/bottles are NOT included and can be ordered separately. C19422 CABKC19422 Each Retrieving

Centrifuge Packages, 200 – 230 V
Description Electrical Includes Supplier No. VWR Catalog Number Unit Price Quantity
Avanti J15R IVD Fixed Angle Package 200 – 230 V Includes benchtop centrifuge and BioCertified JA-10.100 (6 x 100 mL) fixed angle rotor. Adapters, tubes/bottles are NOT included and can be ordered separately C19390 CABKC19390 Each Retrieving
Avanti J15R Swinging Bucket 200 – 230 V Includes benchtop centrifuge and BioCertified JA-10.100 (6 x 100 mL) fixed angle rotor. Adapters, tubes/bottles are NOT included and can be ordered separately C19395 CABKC19395 Each Retrieving
Avanti J15R IVD Swinging Bucket 200 – 230 V Includes benchtop centrifuge and BioCertified JA-10.100 (6 x 100 mL) fixed angle rotor. Adapters, tubes/bottles are NOT included and can be ordered separately C19396 CABKC19396 Each Retrieving
Avanti J-15R, Refrigerated, Benchtop Centrifuge Microplate Package 200 – 230 V Includes Avanti J-15R, 200 – 230 V, 50/60 Hz, refrigerated benchtop centrifuge with LCD touchscreen and the microplate rotor. The JS-4.750u microplate swinging bucket rotor can achieve a maximum speed of 4,450 RPM and a maximum RCF of 4,060xg C19401 CABKC19401 Each Retrieving
Avanti J-15R Cell Culture Package 200 – 230 V Includes Avanti J-15R, 200-230 V, 50/60 Hz,the JS-4.750 rotor, four 15mL conical adapters and four 50mL conical adapters. The swinging bucket rotor can achieves a maximum speed of 4,750 RPM and maximum RCF of 5,250 xg C19413 CABKC19413 Each Retrieving
Avanti J-15R IVD Cell Culture Package 200 – 230 V Includes Avanti J-15R IVD, 200-230 V, 50/60 Hz, the JS-4.750 rotor, four 15mL conical adapters and four 50 mL conical adapters. The swinging bucket rotor can achieve a maximum speed of 4,750 RPM and maximum RCF of 5,250 xg C19414 CABKC19414 Each Retrieving
Avanti J15R Blood Sample Package 200 – 230 V Includes Avanti J-15R, 200 – 230 V, 50/60 Hz,the JS-4.750 rotor, four 13 mm tube adapters and four 16 mm tube adapters. The swinging bucket rotor can achieve a maximum speed of 4,750 RPM and maximum RCF of 5,250 xg C19419 CABKC19419 Each Retrieving
Avanti J15R IVD Blood Sample Package 200 – 230 V Includes Avanti J-15R IVD, 200 – 230 V, 50/60 Hz, the JS-4.750 rotor, four 13 mm tube adapters and four 16 mm tube adapters. The swinging bucket rotor can achieve a maximum speed of 4,750 RPM and maximum RCF of 5,250 xg C19420 CABKC19420 Each Retrieving

Centrifuge Packages, 220 – 230 V
Description Electrical Includes Supplier No. VWR Catalog Number Unit Price Quantity
Avanti J-15R, IVD,Refrigerated, Benchtop Centrifuge Microplate Package 220 – 230 V Includes Avanti J-15R IVD, 200-230 V, 50/60 Hz, refrigerated benchtop centrifuge with LCD touchscreen and the microplate rotor. The JS-4.750 microplate swinging bucket rotor, maximum speed of 4,450 RPM and a maximum RCF of 4,060xg C19402 CABKC19402 Each Retrieving
Avanti J15R Biosafe Package 220 – 230 V Includes Avanti J-15R, 200-230V, 50/60 Hz, the JS-4.750 rotor, 4 aerosolve canisters, 4 15mL conical Biosafe Aerosolve Canister Tube Racks and 4 50mL conical Biosafe Aerosolve Canister Tube Racks, swinging bucket rotor, maximum speed of 4,750RPM C19407 CABKC19407 Each Retrieving
Avanti J15R Ivd Biosafe Package 220 – 230 V Includes Avanti J-15R IVD, 200-230 V, 50/60 Hz, the JS-4.750 rotor, 4 aerosolve canisters, 4 15mL conical Biosafe Aerosolve Canister Tube Racks and 4 50mL conical Biosafe Aerosolve Canister Tube Racks. Swinging rotor, maximum speed 4,750RPM C19408 CABKC19408 Each Retrieving

Centrifuge Packages, 230 V
Description Electrical Includes Supplier No. VWR Catalog Number Unit Price Quantity
Avanti J15R Fixed Angle Package 230 V Includes benchtop centrifuge and BioCertified JA-10.100 (6 x 100 mL) fixed angle rotor. Adapters, tubes/bottles are NOT included and can be ordered separately C19389 CABKC19389 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Model J-15 Ventilated Benchtop Centrifuge J-15R Refrigerated Benchtop Centrifuge
Max. capacity 3 L
Max. speed With JA-10.100 fixed angle rotor: 10,200 rpm;
Using JS-4.750 swinging bucket with tube-and-bottle buckets: 4,550 rpm;
Using multi-well plate carriers (with 1 to 3 stacked plates): 4,350 rpm*
With JA-10.100 fixed angle rotor: 10,200 rpm;
Using JS-4.750 swinging bucket with tube-and-bottle buckets: 4,550 rpm;
Using multi-well plate carriers (with 1 to 3 stacked plates): 4,450 rpm**
Max. RCF (×g) Max. RCF (at rmax) with JA-10.100 fixed angle rotor: 11,420 xg;
Max. RCF (at rmax) using JS-4.750 swinging bucket with tube-and-bottle buckets: 4,820 xg;
Using multi-well plate carriers (1 to 3 stacked plates): 3,880 xg
Max. RCF (at rmax) with JA-10.100 fixed angle rotor: 11,420 xg;
Max. RCF (at rmax) using JS-4.750 swinging bucket with tube-and-bottle buckets: 4,820 xg;
Using multi-well plate carriers (with 1 to 3 stacked plates): 4,060 xg
Width 21.9" (55.6 cm) 29.8" (75.6 cm)
Height 14.5" (36.8 cm)
Depth 29.5" (74.9 cm) 27.7" (70.3 cm)
Temperature setting range –10...+40 °C in 1 °C increments

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping