CytoFLEX Flow Cytometer, Beckman Coulter

Supplier: Beckman Coulter

C09750 B53017 C09752 B53019 C09751 C09754 B53015 B53016
CA76183-348EA 175362.58 CAD
76330-090 CA76183-348 76330-092 CA76102-558 76330-094 76330-530 CA76102-560 CA76102-562
CytoFLEX Flow Cytometer, Beckman Coulter
Cell Counters and Flow Cytometers Flow Cytometers
This Cytometer is designed to deliver superior performance with ease of installation and operation for research applications.

Simplified system setup, data acquisition, analysis, and export of experimental results are integrated into a complete workflow solution with CytExpert software

The CytoFLEX includes 13 band pass filters which can be repositioned as needed, and it is available with different configurations to provide the ultimate in application flexibility, including optional 96-well Plate Loader. Activate the number of channels needed initially and add channels later as research needs grow.

Additional Specifications:

Violet Side Scatter Resolution: (VSSC) <200 nm
Blue Laser Wavelength: 488 nm; Power: 50 mW; Beam spot size: 5 x 80 μm
Red Laser Wavelength: 638 nm; Power: 50 mW; Beam spot size: 5 x 80 μm
Violet Laser Wavelength: 405 nm; Power: 80 mW; Beam spot size: 5 x 80 μm
Flow Cell dimensions: 420 μm x 180 μm internal diameter
Signal Processing: Fully digital system with 7 decade data display
Carryover: Single Tube Format < 1.0%; Plate Loader Format < 0.5%
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR