Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Succinic acid

SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
 
 
SearchResultCount:"9"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Catalog Number: (97061-122)
Supplier:  VWR
Description:   Biological buffers are useful for cell culture in vitro, enzyme assays and some electrophoretic applications at physiological pH.
SDS
Supplier:  Avantor Performance Materials
Description:   Granular.
SDS
Supplier:  Avantor Performance Materials
Description:   The quality of many BAKER ANALYZED™ Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These solvents are es...
SDS
Supplier:  MilliporeSigma
Description:   Cas Number 110-15-6, Chemical Formula Hoocch2Ch2Cooh,For Synthesis
SDS
Catalog Number: (CA1.00682.0250)
Supplier:  MilliporeSigma
Description:   Succinic acid, CAS number:110-15-6, synonyms:Butanedioic acid, Application:for analysis EMSURE
SDS
Supplier:  Alfa Aesar
Description:   SUCCINIC ACID, 99+% 500GM
Supplier:  TCI America
Description:   CAS Number: 110-15-6
MDL Number: MFCD00002789
Molecular Formula: C4H6O4
Molecular Weight: 118.09
Purity/Analysis Method: >...
SDS
Supplier:  MilliporeSigma
Description:   Succinic acid, Cas number: 110-15-6, Synonyms: Butanedioic acid, cryst., suitable for use as excipient EMPROVE* exp NF,JPE, 5 kg
SDS
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 9
  1  2  Next

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping