G-type iODN (inhibitory ODN) (synthetic) endotoxin free (Bulk)
Supplier: Enzo Life Sciences
746-352-M001
89153-132EA
1177.01
CAD
89153-132
G-type iODN (inhibitory ODN) (synthetic) endotoxin free (Bulk)
G-type iODN (inhibitory ODN)
Inhibitory iODN oligodeoxynucleotide with phosphorothioate backbone. Potent sequence of an inhibitory ODN based on poly-G class I sequences targeting TLR9, but without additional effect on TLR7 and TLR8 signaling, for in vivo use in rodents (50-150µg per injection).
- TLR9 inhibitor