Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

You Searched For:

Molecular sieve A4 (0.4 nm, 4 Å)

12  results were found
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
 
 
SearchResultCount:"12"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  Avantor Performance Materials
Description:   Molecular sieves with minimal or no hazard.
Supplier:  Avantor Performance Materials
Description:   Activated, pellets, ‘BAKER ANALYZED'* Reagent, 8-12 mesh. Lot analysis on label. Packaged in a plastic container. 2.5kg
SDS
Catalog Number: (CAMX1583G-1)

Supplier:  MilliporeSigma
Description:   Beads.
SDS
Supplier:  MilliporeSigma
Description:   MOLECULAR SIEVE 4A 1.5KG
SDS
Catalog Number: (CAMX1583J-1)

Supplier:  MilliporeSigma
Description:   Beads.
SDS
Supplier:  Alfa Aesar
Description:   MOLECULR SIEVS 4A 1-2MMBE 250G
Supplier:  Alfa Aesar
Description:   MOLECULR SIEVS 4A 3-5MMBEA 1KG
Supplier:  Alfa Aesar
Description:   MOLECULAR SIEVES 4A POWDER 1KG
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 12
  1  2  Next