pCMV-Tag Vectors, Agilent Technologies
Supplier: Agilent Technologies
211173 240196 240204 211175 240202 211170 240200 240202 240195 240203 240198 240204 211174 211174 211170 211175 211173 240203 240196 211172 240197 211172 240197 240195 240198 240200
CA95040-734EA 821.6 CAD
CA95040-734 95040-742 CA95040-736 CA95040-850 99900-786 99900-788 99901-030 95040-740 99901-032 99901-028 99900-790 95040-734 CA99900-788 CA95040-740 95040-738 CA99900-784 95040-736 CA99900-786 CA95040-742 95040-850 CA99901-032 CA99901-030 CA99901-028 CA99900-790 99900-784 CA95040-738
pCMV-Tag Vectors, Agilent Technologies
Vectors, Plasmids, Libraries and Inserts
The pCMV-Tag vectors have a single copy of the available epitope tags, while the pCMV-3Tag vectors have three copies of the epitope tag (in a variety of configurations) to prove a stronger signal in immmunological assays.
The small size of these tags ensures that they will have minimal interference with functional analysis studies.
Caution: For Research Use Only. Not for use in diagnostic procedures.