Vitality hrGFP II Mammalian Expression Vectors, Agilent Technologies
Supplier: Agilent Technologies
240157 240143 240145 240145 240144 240157 240144 240143
CA97066-376EA 877.96 CAD
CA97066-376 97066-378 CA97066-378 CA97066-382 97066-376 97066-380 CA97066-380 97066-382
Vitality hrGFP II Mammalian Expression Vectors, Agilent Technologies
Vectors, Plasmids, Libraries and Inserts
Vitality hrGFP II Mammalian Expression Vectors support a variety of expression configurations, thus providing ideal expression options for each specific application.
  • Easy to detect
  • Applications include Western blotting, immunoprecipitation and flow cytometry
This is the next generation of original hrGFP that fluoresces brighter in mammalian cells is less toxic than EGFP, making it easier to use for mammalian expression experiments. Since hrGFP is derived from a different organism than EGFP, the hrGFP antibody does not cross react with the Aequorea victoria GFP variant-derived EGFP.
Caution: For research use only. Not for use in diagnostic procedures.