Hazardous Location Blender Motor, Waring®

Supplier: Waring
BHL120 BHL240
58988-009EA 14310.56 CAD
58988-009 58988-010
Hazardous Location Blender Motor, Waring®
Homogenizers Standard Laboratory Blenders
Waring® blenders to provide optimum safety and reliable performance when working with highly volatile substances.

  • For use with 1-liter, stainless steel container only
  • Natural finished aluminum and steel housing
  • Must be hard-wired via explosion-proof conduit
  • Maximum ambient temperature rating 40 °C
  • Speed: 18,500 rpm (no load)

Heat is generated within motor housings during normal operation. This blender is designed to prevent the electrical arcing and generated heat from igniting ambient atmospheres.

Certifications: UL listed motor for use in Class 1, Group D hazardous locations.

Ordering information: One year warranty.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR