PeproGrow™ EPC (Endothelial Progenitor Cell Media), Peprotech, Inc.
Supplier: PeproTech
700-EPC
10777-622EA 615 CAD
10777-622
PeproGrow™ EPC (Endothelial Progenitor Cell Media), Peprotech, Inc.
Cell Culture Media
PeproTech's PeproGrow™ EPC is an antibiotic-free, antimycotics-free, antifungal-free, and phenol red-free maintenance media for endothelial progenitor cells.
  • Complete Media
  • Antibiotic-Free
  • Antimycotics-Free
  • Antifungal-Free
  • Phenol red-Free
PeproTech offers three separate endothelial cell culture media formulations developed for the in vitro cultivation of: endothelial progenitor cells (EPCs; PeproGrow EPC) derived from bone marrow or peripheral blood; endothelial cells from large vessels (PeproGrow™ MacroV); and endothelial cells from small vessels (PeproGrow™ MicroV). These media formulations maintain outstanding endothelial cell morphology and function, and increase the activity of endothelial nitric oxide synthase (eNOS), which account for a specific, crucial marker for endothelial cells. By doing this, the media provide an optimal cell culture environment for macrovascular and microvascular endothelial cells, as well as for EPCs; growing cells at rates that exceed commercially available media.
PeproTech’s endothelial cell culture media kit is supplied as a 500mL bottle of basal medium and a separate growth supplement bottle that contains various essential growth factors and components for endothelial cell growth. Adding the growth supplement to the basal medium results in the complete culture medium. PeproTech’s endothelial media do not contain antibiotics, antimycotics, antifungals, or phenol red, as these components can cause cell stress and masking effects that may reduce complete medium shelf life and influence experimental results.
Caution: PeproGrow™ Endothelial Media Products should be prepared under strict sterile conditions, and stored in the dark to prevent light damaging effects. These products are not intended for use in humans.