Murine IP-10 (CXCL10) Standard ABTS ELISA Development Kit, PeproTech, Inc.
Supplier: PeproTech
900-M153 900-K153
10777-872KT 199.32 CAD
10777-872 10771-584
Murine IP-10 (CXCL10) Standard ABTS ELISA Development Kit, PeproTech, Inc.
Assays ELISAs
PeproTech's Murine IP-10 (CXCL10) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant murine IP-10 (CXCL10) in a sandwich ELISA format.
  • Easy-to-use
  • Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Murine IP-10 (CXCL10) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant murine IP-10 (CXCL10) in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay murine IP-10 (CXCL10) in approximately 1000 ELISA plate wells.