Human IGF-BP1 Standard ABTS ELISA Development Kit, PeproTech, Inc.
Supplier: PeproTech
900-K315 900-M315
10777-726KT 605.54 CAD
10777-726 10781-668
Human IGF-BP1 Standard ABTS ELISA Development Kit, PeproTech, Inc.
Assays ELISAs
PeproTech's Human IGF-BP1 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant hIGF-BP1 in a sandwich ELISA format.
  • Easy-to-use
  • Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Human IGF-BP1 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant hIGF-BP1 in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay hIGF-BP1 in approximately 1000 ELISA plate wells.