Anti-IL-1beta Goat Polyclonal Antibody

Supplier: PeproTech

500-P21BG-50UG 500-P21BG-1MG 500P21BG100UG
10771-184EA 399.75 CAD
10771-184 10771-188 10771-186
Anti-IL-1beta Goat Polyclonal Antibody
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human IL-1β. Anti­Human IL-1β­specific antibody was purified by affinity chromatography employing an immobilized Human IL-1β matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-1beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR