Anti-Leptin Goat Polyclonal Antibody

Supplier: PeproTech
500P185G100UG 500P185G1MG 500P185G50UG
10771-466EA 473.07 CAD
10771-466 10771-468 10771-464
Anti-Leptin Goat Polyclonal Antibody
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Rat Leptin. Anti­Rat Leptin­specific antibody was purified by affinity chromatography employing an immobilized Rat Leptin matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: Leptin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR