Anti-IL-1beta Goat Polyclonal Antibody (Biotin)

Supplier: PeproTech

500P21BGBT50UG 500P21BGBT1MG 500P21BGBT25UG
10771-192EA 512.51 CAD
10771-192 10771-194 10771-190
Anti-IL-1beta Goat Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human IL-1β. Anti­Human IL-1β­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: IL-1beta
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR