Anti-EMAP-II Goat Polyclonal Antibody (Biotin)

Supplier: PeproTech
500P172GBT25UG 500P172GBT50UG 500P172GBT1MG
10776-592EA 369 CAD
10776-592 10776-594 10776-596
Anti-EMAP-II Goat Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human EMAP-II. Anti­Human EMAP-II­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: EMAP-II
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR