Anti-BD-3 Rabbit Polyclonal Antibody (Biotin)

Supplier: PeproTech

500P241BT1MG 500P241BT25UG 500P241BT50UG
10777-168EA 5974.29 CAD
10777-168 10777-164 10777-166
Anti-BD-3 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human BD-3. Anti­Human BD-3­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: BD-3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR