Anti-Vaspin Rabbit Polyclonal Antibody (Biotin)

Supplier: PeproTech

500P256BT25UG 500P256BT50UG 500P256BT1MG
10777-316EA 369 CAD
10777-316 10777-318 10777-320
Anti-Vaspin Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human Vaspin. Anti­Human Vaspin­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: Vaspin
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR