Lambda gt11 Vector Kits, Agilent Technologies
Supplier: Agilent Technologies
234612 234612 234211 234211
CA99900-868EA 1649.76 CAD
99900-868 CA99900-868 99900-866 CA99900-866
Lambda gt11 Vector Kits, Agilent Technologies
Vectors, Plasmids, Libraries and Inserts
For use in the construction of cDNA libraries.
  • Predigested with EcoR I
  • >1×107 pfu/μg lambda DNA must be obtained with test insert (background <1.5%)
These Lambda gt11 vector kits do not allow in vivo excision of cloned fragments. Libraries may be screened with both antibodies and nucleic acid probes.
Caution: For research use only. Not for use in diagnostic procedures.