SuperCos I Vector, Agilent Technologies
Supplier: Agilent Technologies
251301 251301
CA99901-086EA 839.92 CAD
99901-086 CA99901-086
SuperCos I Vector, Agilent Technologies
Vectors, Plasmids, Libraries and Inserts
SuperCos 1 is a novel, 7.9 kb cosmid vector that contains bacteriophage promoter sequences flanking a unique cloning site.
  • Cloning capacity of 30 to 42 kb
  • For generating cosmid libraries
  • Ideal for high-resolution restriction mapping and rapid chromosomal walking
This structure allows rapid synthesis of "walking" probes specific for the extreme ends of insert DNA.
The SuperCos 1 vector is also engineered to contain genes for the amplification and expression of cosmid clones in eukaryotic cells.
Caution: For research use only. Not for use in diagnostic procedures.