Peptide Cleanup Pipette Tips, Agilent Technologies
Supplier: Agilent Technologies
5188-5239 5188-5239
CA76204-912PK 285.57 CAD
CA76204-912 76204-912
Peptide Cleanup Pipette Tips, Agilent Technologies
Pipette Tips
Peptide cleanup pipette tips are packed with ZORBAX SB-C18 silica used for purification and concentration of peptide, protein digests, proteins, or oligonucleotide samples by pipetting sample through the tip. Typical applications include sample preparation for analysis using ESI and MALDI-MS.
  • C18-silica solid phase extraction material is embedded into the wall of the pipette tip (no glue or polymer matrix is used)
  • Maximizing surface area in contact with the sample and providing excellent flow properties
  • Uses an easy-to-follow protocol that provides highly reproducible recoveries of peptides from in-gel digests with low interferences for mass spectrometry (MS) analysis
Recommended one-time use only. Designed to fit all standard 10 µl adjustable pipettes.
Caution: Research use only.