METTLER TOLEDO® Excellence Benchtop Density Meters, Models D4, D5 and D6

Supplier: Mettler-Toledo
30474901 30474902 30474900
76300-948EA 29019.47 CAD
76300-948 76300-950 76300-952
METTLER TOLEDO® Excellence Benchtop Density Meters, Models D4, D5 and D6
Density Meters
Developed for a wide range of applications, the Excellence Density Meters are the next generation of density measurement.

  • Color touchscreen with 7" display
  • Compatible with nine languages
  • Accuracy up to six digits
  • One Click user interface
  • BubbleCheck automatic viscosity correction and temperature control
  • Simultaneous determination of density, refractive index, pH / conductivity and color

The new space-saving Excellence Density line guarantees high reliability and maximum precision. Measure density, specific gravity, as well as many related values such as % alcohol and Brix quickly and accurately.

Ordering information: Can be further enhanced with the following Optional modules and accessories to fit your process needs: powerful autosamplers, LabX PC software, ErgoSens infrared motion sensor, LevelSens waste sensor, LogStraight fingerprint reader, SPR200 sampling pump. Contact your VWR representative for more information.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR