Labconco® Purifier® Rebel Logic®+ Biosafety Cabinets, Labconco
Supplier: Labconco
302310501 302510541 302611511 302610571 302411571 302310571 302411551 302510501 302410541 302610551 302511551 302411521 302310551 302611571 302610511 302610541 302311521 302311541 302310511 302510551 302410511 302510511 302410551 302410571 302510521 302510531 302410521 302311501 302611501 302310531 302510571 302511501 302610521 302611541 302511531 302310541 302411501 302611531 302610531 302310521 302611551 302411531 302311531 302410531 302611521 302311571 302411541 302311551 302411511 302511541 302610501 302311511 302511521 302410501 302511511 302511571
76383-330EA 19405.55 CAD
76383-330 76383-366 76383-430 76383-384 76383-412 76383-342 76383-410 76383-358 76383-352 76383-382 76383-424 76383-404 76383-340 76383-440 76383-374 76383-380 76383-390 76383-394 76383-332 76383-368 76383-346 76383-360 76383-354 76383-356 76383-362 76383-364 76383-348 76383-386 76383-428 76383-336 76383-370 76383-414 76383-376 76383-436 76383-420 76383-338 76383-400 76383-434 76383-378 76383-334 76383-438 76383-406 76383-392 76383-350 76383-432 76383-398 76383-408 76383-396 76383-402 76383-422 76383-372 76383-388 76383-418 76383-344 76383-416 76383-426
Labconco® Purifier® Rebel Logic®+ Biosafety Cabinets, Labconco
Fume Hoods and Enclosures Biological and Microbiological Safety Cabinets
This biosafety cabinet is designed specifically for Discover Echo's Rebel microscope.
  • Class II, Type A2 biosafety cabinet
  • Sump support places microscope stage flush with work surface
  • Less arm and back strain with ergonomic design
Rebel Logic+ BSCs have a unique cutout that allows the microscope stage to sit flush with the work surface of the BSC. This creates less strain on your arms and back while you examine samples. You can view the scope’s large tablet screen at eye level, without eyepieces or sash hole cutouts.
Purifier Rebel Logic+ Class II, Type A2 Biosafety Cabinets provide personnel, ISO 5 product and environmental protection from hazardous particulates such as agents that require Biosafety Level 1, 2 or 3 containment. Other applications include work with antineoplastic drugs, genetic material, asbestos and additional substances that generate hazardous airborne particulates.
These biological safety cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through the optional Ventus Canopy Connection. When exhausted, the biosafety cabinets may be used for applications involving minute quantities of volatile toxic chemicals and tracer amounts of radionuclides as an adjunct to microbiological research.
During operation, room air is drawn into the inlet grill to protect the user. The air in the plenum beneath the work surface is a mixture of unfiltered room air and air that has just passed through the work area. This contaminated air is drawn by the blower through the back plenum of the cabinet, where approximately 70% of the air is recirculated through the supply HEPA filter and back over the work area. The balance of the contaminated air is discharged to the environment after passing through the exhaust HEPA filter.
Certifications: ETL listed. CE conformity marking (230 volt models).
Ordering information: Rebel microscope is not included. May be ordered with or without the accessory package. Accessory package includes 254 nm UV lamp, ADA-compliant service fixture(s) with ball-type valve(s), and NSF-Approved Vacu-Pass™ Cord & Cable Portal.
Caution: Units with various voltages are availble and listed below, please select the unit for your use and voltage requirements carefully.