Metal Detectable Bench Scrapers (8"), Remco
Supplier: Remco
6960MD4 6960MD2 6960MD5 6960MD3
76006-858EA 48.93 CAD
76006-858 76006-856 76006-860 76006-862
Metal Detectable Bench Scrapers (8"), Remco
Scrapers
These metal detectable bench scrapers are excellent solutions for bench top scraping.
  • Large scraping blade clears a sizeable amount of material at once
  • Hand grip promotes a secure grip for the user
  • One-piece construction eliminates seams and crevices where bacteria can hide
The semi-ferrous additive in this product allows for use in conjunction with a metal detectable system. Metal detectable plastic is a lightweight and less costly alternative to stainless steel.