Mini Light Meter
Catalog #: 470315-058
Supplier: Avantor
Discontinued
Specifications
- Description:Light Meter
- Cat. no.:470315-058
Specifications
About this item
With this meter you can measure illuminance from fluorescent, metal halide, high-pressure sodium and incandescent sources.
- Sampling rate: 2.5 times per second for digital display
- Measuring light source: fluorescent, metal halide (HID), high-pressure sodium, and incandescent
- Measuring intensities of illumination in lux or foot-candles
- Data hold
- Maximum/average/minimum recall
- Zero adjustment
- Auto power off and disable function
- Auto ranging
Giving units in both lux and foot-candles, the M110 is durable, reliable, and easy to use.
Customers Who Bought This Also Bought