ContactUsLinkComponent
undefined | Cell Culture Media
Catalog #: 76304-218
Supplier:  Avantor
Catalog #: 76304-218
Supplier:  Avantor
Discontinued
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    100 ml
  • Media Format:
    Liquid
  • Cat. no.:
    76304-218
Specifications
About this item
Serum replacement solution, 100X containing media formulation of non-animal derived, chemically defined salts, designed to replace serum in HEK293, HeLa, or A549 cell culture media.
  • Serum-free
  • Animal-free
  • Protein-free
  • Chemically-defined
  • Cell culture media base: Serum replacement solution
Formulations are proprietary to PeproTech.
Delivery: PeproTech's serum replacement solution is supplied as a 100 ml bottle.
Customers Who Bought This Also Bought