Sulphite - S(IV) Standard, for Ion Chromatography, VeriSpec®, Ricca Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010944-250N RV010944-500N
RCRV010944250CEA 180.5 CAD
RCRV010944250C RCRV010944500C
Sulphite - S(IV) Standard, for Ion Chromatography, VeriSpec®, Ricca Chemical Company
Standards Anionic Standards Sulphite Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml