Polygosil 60-5 C8, HPLC Columns
Supplier: VWR
Polygosil
POLY-5C8-150A POLY-5C8-150AS
76402-304EA 1428.84 CAD
76402-304 76402-306
Polygosil 60-5 C8, HPLC Columns
Chromatography Columns
Irregular silica phases for analytical applications.
  • pH stability: 2 to 8