Masterflex® L/S Variable-Speed Console Drives, Cole-Parmer
Supplier: Cole Parmer
07555-00 07555-05
76403-058EA 2241.42 CAD
76403-058 76403-060
Masterflex® L/S Variable-Speed Console Drives, Cole-Parmer
Pumps Peristaltic Pumps
Compact and cost effective for general fluid transfer applications.
  • Flow range: 0.42 to 2900 ml/min
  • Simple operation with single-turn speed control
  • Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
  • Reversible motor lets you easily prime/purge tubing and pump in either direction
  • Remote control of start/stop via DB9 connector on back of drive
Drives accept nine different pump head types: Standard, Easy-Load®, Easy-Load II, Easy-Load 3, Multichannel, High-Performance, Cartridge, PTFE tubing, and PTFE-diaphragm. Drives 07555-00 and -05 are not recommended for use with the PTFE-tubing and PTFE-diaphragm pump heads.
Ordering information: 1.8 m (6 ft.) line cord with IEC 320/CEE22 socket. Shipped with country-specific cord/plug set; please specify ultimate destination when ordering.