Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • Environmentally Preferable (5)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "laboratory-coveralls"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"22"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   Coveralls are manufactured from a specially formulated breathable microporous fabric that is exceptionally clean and durable. These low-linting dispos...
Supplier:  VWR International
Description:   These are the cleanest and most durable coveralls available within the VWR Protection line. Manufactured from a specially formulated breathable microp...
Supplier:  VWR International
Description:   Protective, durable, and comfortable coveralls provide an excellent barrier against dry particulates, nonhazardous liquids, and microorganisms. Garmen...
Supplier:  VWR International
Description:   Coveralls are manufactured from a unique multilayered spunbonded-meltblown-spunbonded (SMS) fabric that offers high tensile strength while remaining s...
Supplier:  VWR International
Description:   Coveralls are manufactured from a spunbonded polypropylene (SPP) fabric that is strong, lightweight, and breathable.
Supplier:  VWR International
Description:   Soft, strong coveralls provide barrier protection against fluids, particulates, and light chemical splash.
Limited Availability
Supplier:  VWR International
Description:   Blue Coveralls are manufactured from a spunbonded polypropylene (SPP) fabric that is strong, lightweight (30g/m²), and breathable.
Supplier:  VWR International
Description:   Coveralls provide high levels of protection and durability while remaining comfortable. A unique flash spinning process creates an excellent barrier a...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 22
  1  2  3  Next