Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • MWBE (7)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Disinfectant Wipes

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"8"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  Kimberly-Clark
Description:   Waterless wash-up is easy with Waterless Cleaning Wipes, particularly for on-the-go maintenance and cleaning tasks
Supplier:  Vileda Professional - FHP
Description:   MicroOne has proven performance of 99.9% bacterial removal

Supplier:  Advantus
Description:   For cleaning and disinfecting telephones, microphones, and other shared communication equipment
Supplier:  Current Technologies
Description:   Low-linting wipes and liners are specifically designed to protect workers and labs from contamination from blood, body fluids, tissue, specimens plus ...
Minority or Woman-Owned Business Enterprise
Catalog Number: (95041-714)

Supplier:  Dukal
Description:   These 2-ply Alcohol Prep Pads are saturated with 70% isopropyl alcohol. Ideal for use as an antiseptic cleanser.
Catalog Number: (CA70000-120)

Supplier:  Cardinal Health
Description:   These moist soap towelettes are ideal for midstream clean-catch urine specimen procedures

Supplier:  Advantus
Description:   Disposable, lint-free pad is presaturated for cleaning optical-grade surfaces
Catalog Number: (58019-522)

Supplier:  Advantus
Description:   Reduces bacteria on hands after contact with contaminated surfaces and materials
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 8
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping