Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Distillation Adapters

https://ca.vwr.com/stibo/smallweb/std.lang.all/51/41/18945141.jpg

Offered in straight or bent delivery formats, the distilling adapters also feed condensed vapor into collecting receivers. The glassware is resistant to high temperatures as well as chemicals to withstand processing with harsh substances. Ventilated styles allow the system to remain somewhat open since heating an entirely closed system could be dangerous. With standard socket and cone sizes, the distilling adapters neatly fit corresponding components in the purification apparatus for clean eluent flows.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"8"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  Wilmad-LabGlass
Description:   These borosilicate glass three-way adapters have a Standard Taper outer joint on top to accommodate a thermometer, bottom Standard Taper inner joint, ...
Supplier:  Wilmad-LabGlass
Description:   With side arm at angle of 75° from lower part of main tube, these borosilicate glass distilling adapters have three standard taper joints of the same ...
Supplier:  Wilmad-LabGlass
Description:   This borosilicate glass three-way adapter has a Standard Taper 10/30 outer joint on top to accommodate a thermometer, bottom inner Standard Taper 24/4...
Supplier:  Wilmad-LabGlass
Description:   Borosilicate glass distilling adapter is used for fractional distillations at reduced pressures.
Supplier:  Wilmad-LabGlass
Description:   Made of borosilicate glass, these Air-Tite adapters are used for evacuation and addition of inert gases.
Supplier:  Wilmad-LabGlass
Description:   With upper Standard Taper outer joint and same size lower Standard Taper inner joint, these borosilicate glass Kjeldahl adapters are placed above a fl...
Supplier:  Wilmad-LabGlass
Description:   Borosilicate glass distilling adapter is used for fractional distillations at reduced pressures.
Catalog Number: (62939-996)

Supplier:  CORNING CA
Description:   These PYREX® 60° angle distilling connecting adapters have a 24/40 Standard Taper outer joint at the upper end and inner 24/40 Standard Taper joints a...
Product available on GSA Advantage®
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 8
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping