Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Sale Items
You Searched For:
Cork Stoppers
8  results were found
https://ca.vwr.com/stibo/smallweb/std.lang.all/50/75/18945075.jpg
Compressed for a fine texture, cork stoppers are breathable container closures that do not compromise the protection of contents. Resistant to pressure, the devices will not pop off even with interior gas built up. Easily cut the environmentally friendly alternative with a metal borer to insert glass tubing. Although choosing the correct diameter size is still important, the raw material of cork stoppers has a natural ability to adapt to the opening shape for a leak-proof seal.
SearchPresentationType-VERTICAL
SearchResultCount:"8"
  List View Easy View
Sort by:
Supplier:  VWR International
Description:   XXXX Corks function as a closure for bottles, vials, laboratory and other openings.
Product and/or packaging contains recycled/renewable content
Catalog Number: (CA23417-000)
Supplier:  VWR International
Description:   XXXX Corks function as a closure for bottles, vials, laboratory and other openings.
Product and/or packaging contains recycled/renewable content
Supplier:  Manton Industrial
Description:   XXXX Quality Corks have a fine grain and are ideal for use as tight-fitting stoppers.
Product and/or packaging contains recycled/renewable content
Supplier:  G.S.C. INTERNATIONAL, INC.
Description:   XX Quality Corks, Size 20
Supplier:  Manton Industrial
Description:   Regular length, XX quality corks in convenient assortments for laboratory use. Packaged with equal amounts of each size.
Product and/or packaging contains recycled/renewable content
Catalog Number: (470104-138)
Supplier:  Manton Industrial
Description:   These natural corks are perfect for laboratory and industrial use! This pack of 100 corks are ideal for closing up your lab bottles and jars.
Product and/or packaging contains recycled/renewable content
Supplier:  United Scientific Supplies
Description:   Cork stoppers function as a closure for bottles, vials, laboratory vessels and other openings.
Catalog Number: (470104-130)
Supplier:  Manton Industrial
Description:   Cork, Assortment, Student
Product and/or packaging contains recycled/renewable content
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address
VWR will not lift restrictions for residential shipping addresses.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 8