Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

PTFE Tubing

https://ca.vwr.com/stibo/smallweb/std.lang.all/96/25/18989625.jpg

PTFE tubing has high chemical resistance, low and high temperature capability, resistance to weathering, low friction, and electrical and thermal insulation. PTFE’s mechanical properties are low compared to other plastics, but they are often enhanced by adding fillers such as glass fibers or carbon. When filled, PTFE tubing maintains its excellent chemical and temperature resistance properties, while fillers improve mechanical strength, stability, and wear resistance. Because of its non-reactive nature, it is often used in containers and pipework for reactive and corrosive chemicals.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"15"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  MilliporeSigma
Description:   Optifix WANDERER flexible PTFE drain tubing and dosing valve with metal spring.
SDS
Supplier:  Bel-Art Products
Description:   Teflon® PTFE plastic tubing can be ordered by the foot, with a minimum cut of 25 feet, to suit a variety of needs.
Supplier:  S.C.A.T
Description:   All tubing comes in 3 m units
Supplier:  DWK Life Sciences (KIMBLE®)
Description:   For HPLC applications. 1.6mm O.D. PTFE tubing. 3m.
Supplier:  S.C.A.T
Description:   All plastic tubes are one meter in length.
Catalog Number: (CA97012-052)
Supplier:  PerkinElmer
Description:   PTFE-HOSE ID=1.0 X WD=0.3
Supplier:  MicroSolv Technology Corp
Description:   Low pressure and solid color PTFE tubings.
Supplier:  DWK Life Sciences (KIMBLE®)
Description:   Tubing nuts and ferrules are used to make connections between the tubing and the end fittings of CHROMAFLEX® columns.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 15
  1  2  Next

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping