Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Metal Bottles

https://ca.vwr.com/stibo/smallweb/std.lang.all/95/17/18989517.jpg

Metal bottles are ideal for storage applications where glass is not applicable due to the risk of breakage or chemical incompatibility. The bottles feature welded seams with rounded inner edges and polished, brushed exteriors. All metal vessels are autoclavable to guarantee that materials stored, transported, or sent are not affected by previous substance residue. Storing completely unaltered, the metal will protect against outside influences such as UV radiation and extreme temperatures.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"4"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  BURKLE INC
Description:   The aluminum bottle is particularly suited for storing, transporting, or sending substances and reference samples which have to be stored completely u...
Catalog Number: (75880-448)
Supplier:  Qorpak
Description:   24 gauge black steel phenolic lined closed head pail with 2" bung and Rieke plug.
Supplier:  Ace Glass
Description:   SCHOTT DURAN® 316L stainless steel bottles are ideal for storage applications where glass is not applicable due to the risk of breakage or chemical in...
Small Business Enterprise
Supplier:  BURKLE INC
Description:   Safety containers for transport and storage are made of high-quality stainless steel AISI 304 (1.4301)/AISI 316 Ti (1.4571). Containers have a uniform...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 4  of 4