Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Sale Items
  • Search Within Results

Powder Funnels

https://ca.vwr.com/stibo/smallweb/std.lang.all/53/37/18945337.jpg

When pouring large particles into reaction vessels, powder funnels allow a clean and quick procedure. Manufactured from clear glass or indestructible plastic, powder funnels have a resistant construction permitting use with various reagents and even chemicals. Powder bridging is minimized by the parallel, short stems. The funnels may have a tapered or angled stem to accommodate neck openings. Featuring ribbing to release air, the disposable or reusable models are offered in various diameter sizes.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"42"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   Polypropylene funnels feature a 60° body angle and 58° internal rib angle for rapid filtration.
Supplier:  VWR International
Description:   Powder funnels are fast and efficient.
Product available on GSA Advantage®
Supplier:  VWR International
Description:   Funnels comes with short, wide stem, and are useful for adding powder chemical to reaction vessels.
UL Listed
Supplier:  VWR International
Description:   Polypropylene funnels are steam autoclavable at 121°C (250°F).

Supplier:  VWR International
Description:   VWR® disposable paper lab funnels are a simple and cost-effective solution to reduce plastic use in the lab and reduce cross-contamination from reuse ...
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Catalog Number: (470307-012)

Supplier:  Heathrow Scientific
Description:   A clean transfer every time.
Supplier:  Thermo Scientific
Description:   Nalgene® Polypropylene Powder Funnels are used for transferring powders
Supplier:  DWK Life Sciences (KIMBLE®)
Description:   Funnels feature short, wide stems and fire-polished tops and stems.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 42
  1  2  3  4  5  6  Next 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping