Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Toluene-α-sulphonyl fluoride

SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
 
 
SearchResultCount:"9"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  Alfa Aesar
Description:   A-TOLUNESULFONYL FLRIDE 99% 5G
Supplier:  Enzo Life Sciences
Description:   Irreversible inhibitor of serine proteases. Widely used fatty acid amid hydrolase (FAAH) inhibitor for pretreatment of brain membrane preparations whe...
Catalog Number: (10187-508)
Supplier:  BioVision
Description:   A serine protease inhibitor, Synonyms: (A)-Toluenesulfonyl fluoride, Benzylsulfonyl fluoride, Phenylmethylsulfonyl fluoride, CAS Number: 329-98-6, Purity: ≥99%, Molecular Formula: C₇H₇FO2S, 5g.
SDS
Catalog Number: (10190-958)
Supplier:  BioVision
Description:   PMSF, A serine protease inhibitor, Alternate Name/Synonyms: A-Toluenesulfonyl fluoride, Benzylsulfonyl fluoride, Phenylmethylsulfonyl fluoride, CAS Number: 329-98-6, Purity: ≥99%, An irreversible inhibitor of serine proteases and mammalian acetylcholinesterase, 25 mg
SDS
Supplier:  Bachem Americas
Description:   Sequence: Phenylmethylsulfonyl fluoride
CAS-No.: 329-98-6
Mol.weight: 174.20
SumFormula: C₇H₇FO₂S
Bulk item no.: Q-1395
Synon...
Supplier:  MilliporeSigma
Description:   White powder. Insoluble in water.
SDS
Supplier:  MilliporeSigma
Description:   Irreversible inhibitor of serine proteases. Its mechanism of action is analogous to that of diisopropylfluorophosphate. PMSF causes sulfonylation of t...
SDS
Catalog Number: (CA82021-256)
Supplier:  G-Biosciences
Description:   Ultra-Pure Individual Protease Inhibitors. Inhibits serine proteases (chymotrypsin, trypsin and thrombin) and cysteine proteases (papain).
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 9
  1  2  Next

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping