4-Methyl-2-pentanone Single-Component Organic Standard, VWR Chemicals BDH®
Supplier: VWR Chemicals BDH®
BDH0700-1ML
BDH0700-1MLEA 153.32 CAD
BDH0700-1ML
4-Methyl-2-pentanone Single-Component Organic Standard, VWR Chemicals BDH®
Standards Organic Standards
Single-Component Organic Standards
They are supplied in methanol, or in another solvent, where appropriate. Additional solvents include methylene chloride, acetone, benzene, hexane, acetonitrile, THF, and water.
Ordering information: Please inquire about the matrix for a particular item when ordering, if the matrix is important to your application, a substitution may be possible.
Concentrations other than these, as well as custom matrices, are readily available, but a minimum order of five ampules may be required in some cases.