AcroPak™ Filtration Capsules, Pall Laboratory

Supplier: Pall Laboratory
AcroPak™
12461 12999 12686 12471 12467 12995 12460 12094 12993 12471 12461 12675 12464 12473 12994 12991 12478 12686 12466 12999 12093 12095 12069 12994 12094 12997 12069 12095 12995 12996 12093 12996 12675 12466 12992 12473 12464 12993 12941 12460 12997 12467 12463 12992 12463 12478 12941 12991
CA97035-170PK 536.35 CAD
97040-968 28143-820 CA97035-170 28143-864 97040-966 CA97035-172 97040-964 87003-890 28143-824 97040-962 28146-178 CA97040-972 28146-179 28143-969 28146-198 CA28146-185 28146-177 28143-828 CA97040-970 CA28146-180 28146-196 CA28146-183 CA28146-182 CA28146-181 CA97040-968 CA97040-966 CA97040-964 CA28143-820 CA28143-864 CA97040-962 CA28143-824 CA28146-177 CA28146-198 CA87003-890 CA28143-969 CA28146-196 97040-972 CA28143-828 97040-970 CA28146-179 CA28146-178 28146-181 28146-182 28146-180 28146-185 28146-183 97035-172 97035-170
AcroPak™ Filtration Capsules, Pall Laboratory
Filters Membrane Filter Capsules
A filter for every step in your process. From drug discovery and basic research to process development and production, Pall Laboratory’s AcroPak capsules come in a variety of membrane types and volume sizes.

  • 100% integrity tested (where noted)
  • Endotoxin level: <0.25 EU/ml using Limulus Amebocyte Lysate test
  • Bacterial retention: 0.2 μm lot samples retain 10⁷ cfu/cm² of B. diminuta per modified ASTM F838
  • Mycoplasma reduction: 0.1 μm lot samples retain 10⁷ cfu/cm² A. Laidlawii per modified ASTM F838
  • Biological safety: Passes United States Pharmacopeia (USP) biological reactivity test in vivo <88>
  • Sterilisation by gamma irradiation

Primary applications include: Buffer media and reagent manufacturing, protein purification and cell culture and bioburden filtration.

Supor PES (Polyethersulfone):
With high flow and capacity, broad fluid compatibility, low protein adsorption, active ingredients, and stabilizers, Pall PES membranes are best suited for filtration of preparative fluids such as buffers, media, and bioprocess intermediates.

Supor EKV Asymmetric PES:
PES filters with asymmetric membrane layers have the same benefits of Supor PES (above), but with additional debris holding. This makes them ideal for high turbidity, high bioburden solutions. All Supor EKV products are a 0.65 µm pre-filter and a 0.2 µm asymmetric membrane.

Fluorodyne II PVDF (hydrophilic polyvinylidene fluoride):
Low extractables, low product and excipient adsorption, simple validation for the sterilization of a wide range of formulations make PVDF membranes ideal for final filtration.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR