J.T.Baker® Robotic Tips Type MP
Catalog #: 49209-0105
Supplier: Ritter, part of Avantor
Price
924,33 $
Log in for availability
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Specifications
- Volume:1000 µl
- Tip Color:Black
- Packed:8× 5 racks, 96 each, blistered
- Cat. no.:49209-0105
- Filter:+
- Sterile:-
Specifications
About this item
Suitable for use with Roche MagNA Pure instruments. Available in conductive version, with filter.
- Manufactured under cleanroom conditions (cleanroom Class 8)
- State of the art quality control
- Barcoded trays for automated identification
- Free of DNA, DNase, RNase and PCR inhibitors
- Ideal for high throughput analysis
- Packed in blisters
The validation of the tips on the instrument for specific applications is within the responsibility of the user.
Customers Who Bought This Also Bought