Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

cotton

https://ca.vwr.com/stibo/smallweb/std.lang.all/50/77/18945077.jpg

Used to efficiently seal tubes or plug pipettes, the cotton acts as a filter allowing air transference while restricting particulates from entering containers. The soft, flexible fiber can easily be manipulated into any container opening. Creating an aerated interior condition, the rolls and balls prevents gas from accumulating inside which could lead to hazardous situations. With absorbent characteristics, the cotton cushions contents from any impact during transport or handling.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"8"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  US Cotton
Description:   Bleached cotton available in sterile and nonsterile. Choose from two types: cotton rolls or cotton balls.
Supplier:  Tidi Products
Description:   Manufactured for maximum absorbency and uniform consistency, shape, and size. Made in the USA.
Catalog Number: (470146-894)
Supplier:  U.S. COTTON, LLC
Description:   COTTON BALLS MEDIUM BX4000
Catalog Number: (CA95041-718)
Supplier:  Dukal
Description:   These cotton balls are made of 100% cotton and are absorbent and clean. They are available in 2 sizes.
Supplier:  US COTTON, LLC
Description:   For General Laboratory Use.
Supplier:  Dukal
Description:   This 100% cotton roll is ideal for cleaning wounds, for application of medications, and where padding is needed. It is easy to unwind, and the short f...
Catalog Number: (470150-780)
Supplier:  US COTTON, LLC
Description:   COTTON 14 G (ROLL 4X68 ) 1/2 OZ.
Supplier:  US Cotton
Description:   These 100% cotton, nonsterile cotton squares feature quilting and interwoven layers for strength. They are packaged with a resealable zip closure for ...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 8